Learn More
Medchemexpress LLC Tam557 Tfa | 98.5% | 1119.34 | 5 MG
SDP
Catalog No. 5000393152
Encompass_Preferred
TAM557 TFA is a cytotoxic tubulysin compound modified for conjugation to transport vehicles such as proteins, peptides, small molecules, or polymeric carriers that can carry a targeting principle. It is for research use only and not sold to patients.
  • Cytotoxic tubulysin compound
  • Modified for conjugation to transport vehicles
  • Available in 1 mg, 5 mg, 10 mg, and 50 mg sizes
Catalog No. 50-003-93152 Supplier Medchemexpress LLC Supplier No. HY160473A5MG
TAM557 TFA is a cytotoxic tubulysin compound modified for conjugation to transport vehicles such as proteins, peptides, small molecules, or polymeric carriers that can carry a targeting principle. It is for research use only and not sold to patients.
  • Cytotoxic tubulysin compound
  • Modified for conjugation to transport vehicles
  • Available in 1 mg, 5 mg, 10 mg, and 50 mg sizes