Learn More
Medchemexpress LLC Tamapin TFA | 98.8% | 3458.11 | 5 MG
SDP
Catalog No. 5000398229
Encompass_Preferred
Tamapin TFA is a venom peptide, targeting to small conductance Ca(2+)-activated K(+) (SK) channels. It is a selective blocker of SK2 (Potassium Channel) and inhibits SK channel-mediated currents in pyramidal neurons of the hippocampus. Tamapin TFA can be isolated from the Indian red scorpion (Mesobuthus tamulus).
  • Targets small conductance Ca(2+)-activated K(+) (SK) channels
  • Selective blocker of SK2 (Potassium Channel)
  • Inhibits SK channel-mediated currents in pyramidal neurons of the hippocampus
Catalog No. 50-003-98229 Supplier Medchemexpress LLC Supplier No. HYP5154A5MG
Tamapin TFA is a venom peptide, targeting to small conductance Ca(2+)-activated K(+) (SK) channels. It is a selective blocker of SK2 (Potassium Channel) and inhibits SK channel-mediated currents in pyramidal neurons of the hippocampus. Tamapin TFA can be isolated from the Indian red scorpion (Mesobuthus tamulus).
  • Targets small conductance Ca(2+)-activated K(+) (SK) channels
  • Selective blocker of SK2 (Potassium Channel)
  • Inhibits SK channel-mediated currents in pyramidal neurons of the hippocampus