Learn More
MOSES LAKE INDUSTRIES INC TAMAPURE-AA-100 H2O2 35%, Ultra-High Purity, Trace Analysis Blank Values, 500 g
Catalog No. NC3593151
Encompass_Preferred
TAMAPURE-AA-100 Hydrogen Peroxide (H2O2) 35%, 500 g, offers ultra-high purity for analytical procedures. Its low metallic impurity levels (<100 ppt) enhance contamination control, improving analytical accuracy. This reagent is specifically designed for sample pretreatment in research applications, ensuring high consistency across lots due to rigorous quality controls. Ideal for environments meeting CLASS 100 clean standards, it is an essential choice for laboratories seeking reliable performance in critical research processes.
Catalog No. NC3593151 Supplier MOSES LAKE INDUSTRIES INC Supplier No. AA10HYDRO PEROX
TAMAPURE-AA-100 Hydrogen Peroxide (H2O2) 35%, 500 g, offers ultra-high purity for analytical procedures. Its low metallic impurity levels (<100 ppt) enhance contamination control, improving analytical accuracy. This reagent is specifically designed for sample pretreatment in research applications, ensuring high consistency across lots due to rigorous quality controls. Ideal for environments meeting CLASS 100 clean standards, it is an essential choice for laboratories seeking reliable performance in critical research processes.