Learn More
Selleck Chemical LLC Tamarixetin E2502-5MG
SDP
Catalog No. 5000039885
Encompass_Preferred
Tamarixetin (4 -O-Methyl Quercetin) is a natural flavonoid derivative of quercetin and has superior anti-inflammatory properties in bacterial sepsis by increasing the population of IL-10-secreting immune cells
Catalog No. 50-000-39885 Supplier Selleck Chemical LLC Supplier No. E25025MG
Tamarixetin (4 -O-Methyl Quercetin) is a natural flavonoid derivative of quercetin and has superior anti-inflammatory properties in bacterial sepsis by increasing the population of IL-10-secreting immune cells