Learn More
TARGETMOL CHEMICALS INC TAMARIXETIN 5MG
SDP
Catalog No. 502578899
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tamarixetin (4'-O-Methyl Quercetin) (4'-O-Methyl Quercetin) is a natural flavonoid derivative of quercetin with anti-inflammatory and anti-oxidative effects. Tamarixetin protects against cardiac hypertrophy. purity: 99%
Catalog No. 50-257-8899 Supplier TARGETMOL CHEMICALS INC Supplier No. TN10395MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tamarixetin (4'-O-Methyl Quercetin) (4'-O-Methyl Quercetin) is a natural flavonoid derivative of quercetin with anti-inflammatory and anti-oxidative effects. Tamarixetin protects against cardiac hypertrophy. purity: 99%