Learn More
US Plastic Corporation Tamco® Tray, 10-3/8" L x 10-3/8" W x 1-1/2" Hgt. Green
SDP
Catalog No. NC3845280
Encompass
These heavy-duty Tamco® Trays are durable, strong, and great for use in a variety of industrial applications. Made of a UV stabilized LLDPEChemical resistantWall thickness: 3/16"Withstands temperature up to 140°FMeets FDA standardsDimensions are nominal
Catalog No. NC3845280 Supplier US Plastic Corporation Supplier No. 15353
These heavy-duty Tamco® Trays are durable, strong, and great for use in a variety of industrial applications. Made of a UV stabilized LLDPEChemical resistantWall thickness: 3/16"Withstands temperature up to 140°FMeets FDA standardsDimensions are nominal