Learn More
US Plastic Corporation TAMCO TRAY 12X16X6IN
SDP
Catalog No. NC2390293
Encompass
Welded High Density Polyethylene constructionWall thickness: 3/16"Rigid, sturdy constructionCorrosion resistantDimensions are inside dimensionsMaximum temperature: 140°FMaterials meet FDA standards
Catalog No. NC2390293 Supplier US Plastic Corporation Supplier No. 14933
Welded High Density Polyethylene constructionWall thickness: 3/16"Rigid, sturdy constructionCorrosion resistantDimensions are inside dimensionsMaximum temperature: 140°FMaterials meet FDA standards