Learn More
US Plastic Corporation Tamco® Tray, 30" L x 24" x 3" Hgt. Red
SDP
Catalog No. NC3845531
Encompass
Available in an array of sizes, these heavy-duty Tamco® Trays are durable, strong, and great for use in a variety of industrial applications.Made of a UV stabilized LLDPEChemical resistantWall thickness: 3/16"Withstands temperature up to 140°FMeets FDA standardsDimensions are nominal
Catalog No. NC3845531 Supplier US Plastic Corporation Supplier No. 17948
Available in an array of sizes, these heavy-duty Tamco® Trays are durable, strong, and great for use in a variety of industrial applications.Made of a UV stabilized LLDPEChemical resistantWall thickness: 3/16"Withstands temperature up to 140°FMeets FDA standardsDimensions are nominal