Learn More
US Plastic Corporation 30" L x 12" W x 3" Hgt. Natural Tamco® Tray
SDP
Catalog No. NC3734728
Encompass
Tamco® Trays are durable, strong, and great for use in a variety of industrial applications.-Made of a UV stabilized LLDPE-Chemical resistant-Wall thickness: 3/16"-Withstands temperature up to 140°F-Meets FDA standards-Dimensions are nominal
Catalog No. NC3734728 Supplier US Plastic Corporation Supplier No. 18626
Tamco® Trays are durable, strong, and great for use in a variety of industrial applications.-Made of a UV stabilized LLDPE-Chemical resistant-Wall thickness: 3/16"-Withstands temperature up to 140°F-Meets FDA standards-Dimensions are nominal