Learn More
Medchemexpress LLC HY-13255 50mg Medchemexpress, TAME CAS:901-47-3 Purity:>98%
SDP
Catalog No. 502081316
Encompass_Preferred
Medchemexpress, HY-13255 50mg TAME CAS:901-47-3 TAME is an inhibitor of anaphase-promoting complex/cyclosome (APC/C or APC), which binds to APC/C and prevents its activation by Cdc20 and Cdh1, produces mitotic arrest. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1316 Supplier Medchemexpress LLC Supplier No. HY1325550MG
Medchemexpress, HY-13255 50mg TAME CAS:901-47-3 TAME is an inhibitor of anaphase-promoting complex/cyclosome (APC/C or APC), which binds to APC/C and prevents its activation by Cdc20 and Cdh1, produces mitotic arrest. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.