Learn More
Medchemexpress LLC HY-14652 10mg Medchemexpress, Tamibarotene CAS:94497-51-5 Purity:>98%
SDP
Catalog No. 502030550
Encompass_Preferred
Medchemexpress, HY-14652 10mg Tamibarotene CAS:94497-51-5 Tamibarotene is a retinoic acid receptor α/β (RARα/β) agonist, showing high selectivity over RARγ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0550 Supplier Medchemexpress LLC Supplier No. HY1465210MG
Medchemexpress, HY-14652 10mg Tamibarotene CAS:94497-51-5 Tamibarotene is a retinoic acid receptor α/β (RARα/β) agonist, showing high selectivity over RARγ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.