Learn More
TARGETMOL CHEMICALS INC TAMIBAROTENE 200MG
SDP
Catalog No. 502579304
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tamibarotene (Amnolake) is an orally active synthetic retinoid developed to overcome all-trans retinoic acid (ATRA) resistance with potential antineoplastic activity. purity: 98%
Catalog No. 50-257-9304 Supplier TARGETMOL CHEMICALS INC Supplier No. T6694200MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tamibarotene (Amnolake) is an orally active synthetic retinoid developed to overcome all-trans retinoic acid (ATRA) resistance with potential antineoplastic activity. purity: 98%