Learn More
Medchemexpress LLC Tamibarotene | 94497-51-5 | 99.57% | 351.44 | 200 MG
SDP
Catalog No. 5000363902
Encompass_Preferred
Tamibarotene is an orally active retinoic acid receptor α (RARα) agonist, demonstrating high selectivity over RARγ. It is intended for research use only.
  • Orally active retinoic acid receptor α (RARα) agonist
  • High selectivity over RARγ
  • Suitable for research use only
Catalog No. 50-003-63902 Supplier Medchemexpress LLC Supplier No. HY14652200MG
Tamibarotene is an orally active retinoic acid receptor α (RARα) agonist, demonstrating high selectivity over RARγ. It is intended for research use only.
  • Orally active retinoic acid receptor α (RARα) agonist
  • High selectivity over RARγ
  • Suitable for research use only