Learn More
Medchemexpress LLC Tamnorzatinib 5mg | 1646839-59-9 | 5 MG
SDP
Catalog No. 5000229465
Encompass_Preferred
Tamnorzatinib (ONO-7475) is a small-molecule Axl and Mer (MERTK) receptor tyrosine kinase inhibitor supplied as a solid for laboratory research. It is intended for in vitro biochemical assays and cellular studies and is not for human therapeutic use.
  • Potent inhibition of Axl and Mer kinases.
  • High chemical purity suitable for research applications.
  • Available in small solid quantities for assay setup.
  • Soluble in common organic solvents, for stock solution preparation.
Catalog No. 50-002-29465 Supplier Medchemexpress LLC Supplier No. HY1143585MG
Tamnorzatinib (ONO-7475) is a small-molecule Axl and Mer (MERTK) receptor tyrosine kinase inhibitor supplied as a solid for laboratory research. It is intended for in vitro biochemical assays and cellular studies and is not for human therapeutic use.
  • Potent inhibition of Axl and Mer kinases.
  • High chemical purity suitable for research applications.
  • Available in small solid quantities for assay setup.
  • Soluble in common organic solvents, for stock solution preparation.