Learn More
Medchemexpress LLC Tamoxifen | 10540-29-1 | 99.96% | 371.51 | 200 MG
SDP
Catalog No. 5000350350
Encompass_Preferred
Tamoxifen is an orally active, selective estrogen receptor modulator (SERM) that blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells. It is a potent Hsp90 activator, enhancing the Hsp90 molecular chaperone ATPase activity. It activates autophagy and induces apoptosis. Tamoxifen can also be dissolved in corn oil for use in inducing gene knockout in CreER transgenic mice.
  • Orally active, selective estrogen receptor modulator (SERM)
  • Blocks estrogen action in breast cells
  • Can activate estrogen activity in bone, liver, and uterine cells
  • Potent Hsp90 activator
  • Enhances Hsp90 molecular chaperone ATPase activity
  • Inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 values of 0.1 μM and 1.8 μM
  • Activates autophagy and induces apoptosis
  • Can be dissolved in corn oil for gene knockout in CreER transgenic mice
Catalog No. 50-003-50350 Supplier Medchemexpress LLC Supplier No. HY13757A200MG
Tamoxifen is an orally active, selective estrogen receptor modulator (SERM) that blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells. It is a potent Hsp90 activator, enhancing the Hsp90 molecular chaperone ATPase activity. It activates autophagy and induces apoptosis. Tamoxifen can also be dissolved in corn oil for use in inducing gene knockout in CreER transgenic mice.
  • Orally active, selective estrogen receptor modulator (SERM)
  • Blocks estrogen action in breast cells
  • Can activate estrogen activity in bone, liver, and uterine cells
  • Potent Hsp90 activator
  • Enhances Hsp90 molecular chaperone ATPase activity
  • Inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 values of 0.1 μM and 1.8 μM
  • Activates autophagy and induces apoptosis
  • Can be dissolved in corn oil for gene knockout in CreER transgenic mice