Learn More
Medchemexpress LLC Tamoxifen | 10540-29-1 | MFCD00010454 | 99.96% | 371.51 | 250mg
SDP
Catalog No. 5000350158
Encompass_Preferred
Tamoxifen (ICI 47699) is an orally active, selective estrogen receptor modulator (SERM) that blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells. It is a potent Hsp90 activator and enhances Hsp90 molecular chaperone ATPase activity.
  • Potently inhibits infectious EBOV Zaire and Marburg (MARV).
  • Activates autophagy and induces apoptosis.
  • Can be dissolved in corn oil for inducing gene knockout in CreER transgenic mice.
  • Has better solubility in corn oil compared to Tamoxifen Citrate.
Catalog No. 50-003-50158 Supplier Medchemexpress LLC Supplier No. 250MGHY13757AR
Tamoxifen (ICI 47699) is an orally active, selective estrogen receptor modulator (SERM) that blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells. It is a potent Hsp90 activator and enhances Hsp90 molecular chaperone ATPase activity.
  • Potently inhibits infectious EBOV Zaire and Marburg (MARV).
  • Activates autophagy and induces apoptosis.
  • Can be dissolved in corn oil for inducing gene knockout in CreER transgenic mice.
  • Has better solubility in corn oil compared to Tamoxifen Citrate.