Learn More
Cayman Chemical Tamoxifen, 10540-29-1, 10 g
SDP
Supplier:  Cayman Chemical 1325810
Encompass_Preferred
Molecular Formula C26H29NO, Purity ≥95%, Formula Weight 371.5; A selective estrogen receptor (ER) modulator; an ER antagonist in breast tissue, effective in treating early breast cancer; an ER agonist in bone and blood vessels and a partial ER agonist in uterine tissues.
Catalog No. NC0601170