Learn More
Medchemexpress LLC HY-13757A 1g Medchemexpress, Tamoxifen CAS:10540-29-1 Purity:>98%
SDP
Catalog No. 502029027
Encompass_Preferred
Medchemexpress, HY-13757A 1g Tamoxifen CAS:10540-29-1 Tamoxifen (ICI 47699) is an orally active, selective estrogen receptor modulator (SERM) which blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells[2][3]. Tamoxifen is a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity. Tamoxifen also potent inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 of 0.1 µM and 1.8 µM, respectively[5]. Tamoxifen activates autophagy and induces apoptosis[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9027 Supplier Medchemexpress LLC Supplier No. HY13757A1G
Medchemexpress, HY-13757A 1g Tamoxifen CAS:10540-29-1 Tamoxifen (ICI 47699) is an orally active, selective estrogen receptor modulator (SERM) which blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells[2][3]. Tamoxifen is a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity. Tamoxifen also potent inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 of 0.1 µM and 1.8 µM, respectively[5]. Tamoxifen activates autophagy and induces apoptosis[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.