Learn More
Medchemexpress LLC HY-13757A 5g , Tamoxifen ICI47699;Z-Tamoxifen;trans-Tamoxifen CAS:10540-29-1 Purity:98%
SDP
Catalog No. 501492171
Encompass_Preferred
Medchemexpress, HY-13757A 5g Tamoxifen ICI47699;Z-Tamoxifen;trans-Tamoxifen CAS:10540-29-1 Formula:C26H29NO Estrogen receptor Purity:98% Tamoxifen is an antiestrogen by inhibiting the binding of estrogen to estrogen receptors . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2171 Supplier Medchemexpress LLC Supplier No. HY13757A5G
Medchemexpress, HY-13757A 5g Tamoxifen ICI47699;Z-Tamoxifen;trans-Tamoxifen CAS:10540-29-1 Formula:C26H29NO Estrogen receptor Purity:98% Tamoxifen is an antiestrogen by inhibiting the binding of estrogen to estrogen receptors . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.