Learn More
Sigma Aldrich Fine Chemicals Biosciences Tamoxifen, 10540-29-1, MFCD00010454, 1g
Catalog No. 501656821
Encompass
Linear Formula C6H5C(C2H5)=C(C6H5)C6H4OCH2CH2N(CH3)2, ≥99%, Molecular Weight 371.51, mp 97-98 °C (lit.), Synonym: (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene, trans-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethylamine
Catalog No. 50-165-6821 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T56481G
Linear Formula C6H5C(C2H5)=C(C6H5)C6H4OCH2CH2N(CH3)2, ≥99%, Molecular Weight 371.51, mp 97-98 °C (lit.), Synonym: (Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene, trans-2-[4-(1,2-Diphenyl-1-butenyl)phenoxy]-N,N-dimethylethylamine