Learn More
Selleck Chemical LLC Tamoxifen Citrate S1972-250mg
SDP
Catalog No. 503109796
Encompass_Preferred
Tamoxifen Citrate is a selective estrogen receptor modulator (SERM) Tamoxifen Citrate is also a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity Tamoxifen induces apoptosis and autophagy
Catalog No. 50-310-9796 Supplier Selleck Chemical LLC Supplier No. S1972250MG
Tamoxifen Citrate is a selective estrogen receptor modulator (SERM) Tamoxifen Citrate is also a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity Tamoxifen induces apoptosis and autophagy