Learn More
Medchemexpress LLC Tamoxifen Citrate | 54965-24-1 | MFCD00058321 | 99.9% | 563.6 g/mol | 200 MG
SDP
Catalog No. 5000352014
Encompass_Preferred
Tamoxifen Citrate is an orally active, selective estrogen receptor modulator (SERM) that blocks estrogen action in breast cells and can activate estrogen activity in other cells. It also acts as a potent Hsp90 activator and potently inhibits infectious EBOV Zaire and Marburg (MARV).
  • Activates autophagy
  • Induces apoptosis
  • Can be used to induce gene knockout in CreER transgenic mice
  • Targets estrogen receptor and HSP90
Catalog No. 50-003-52014 Supplier Medchemexpress LLC Supplier No. HY13757200MG
Tamoxifen Citrate is an orally active, selective estrogen receptor modulator (SERM) that blocks estrogen action in breast cells and can activate estrogen activity in other cells. It also acts as a potent Hsp90 activator and potently inhibits infectious EBOV Zaire and Marburg (MARV).
  • Activates autophagy
  • Induces apoptosis
  • Can be used to induce gene knockout in CreER transgenic mice
  • Targets estrogen receptor and HSP90