Learn More
Medchemexpress LLC HY-13757 5g Medchemexpress, Tamoxifen (Citrate) CAS:54965-24-1 Purity:>98%
SDP
Catalog No. 502029026
Encompass_Preferred
Medchemexpress, HY-13757 5g Tamoxifen (Citrate) CAS:54965-24-1 Tamoxifen Citrate (ICI 46474) is an orally active, selective estrogen receptor modulator (SERM) which blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells[2][3].Tamoxifen Citrate is a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity. Tamoxifen Citrate also potent inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 of 0.1 µM and 1.8 µM, respectively[5]. Tamoxifen Citrate activates autophagy and induces apoptosis[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9026 Supplier Medchemexpress LLC Supplier No. HY137575G
Medchemexpress, HY-13757 5g Tamoxifen (Citrate) CAS:54965-24-1 Tamoxifen Citrate (ICI 46474) is an orally active, selective estrogen receptor modulator (SERM) which blocks estrogen action in breast cells and can activate estrogen activity in other cells, such as bone, liver, and uterine cells[2][3].Tamoxifen Citrate is a potent Hsp90 activator and enhances the Hsp90 molecular chaperone ATPase activity. Tamoxifen Citrate also potent inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 of 0.1 µM and 1.8 µM, respectively[5]. Tamoxifen Citrate activates autophagy and induces apoptosis[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.