Learn More
Apexbio Technology LLC Tamoxifen Citrate 54965-24-1 5g
SDP
Catalog No. 502240249
Encompass_Preferred
Tamoxifen Citrate (CAS 54965-24-1) is a small-molecule antagonist targeting estrogen receptors (ER) It is designed to competitively bind ER thereby modulating estrogen-mediated signal transduction Tamoxifen Citrate exerts its biological activity primarily by inhibiting the transcriptional activity of estrogen-responsive genes leading to reduced synthesis of factors related to cell growth and angiogenesis In in vitro studies Tamoxifen Citrate demonstrates inhibition of cell proliferation with reported IC50 values typically ranging from 3 to 7 M depending on cell line and experimental conditions Based on these pharmacological properties Tamoxifen Citrate holds research potential in studies of estrogen receptor-dependent signaling pathways and anti-tumor strategies particularly in breast cancer models
This item is not returnable. View return policy
Catalog No. 50-224-0249 Supplier Apexbio Technology LLC Supplier No. B13945000
Tamoxifen Citrate (CAS 54965-24-1) is a small-molecule antagonist targeting estrogen receptors (ER) It is designed to competitively bind ER thereby modulating estrogen-mediated signal transduction Tamoxifen Citrate exerts its biological activity primarily by inhibiting the transcriptional activity of estrogen-responsive genes leading to reduced synthesis of factors related to cell growth and angiogenesis In in vitro studies Tamoxifen Citrate demonstrates inhibition of cell proliferation with reported IC50 values typically ranging from 3 to 7 M depending on cell line and experimental conditions Based on these pharmacological properties Tamoxifen Citrate holds research potential in studies of estrogen receptor-dependent signaling pathways and anti-tumor strategies particularly in breast cancer models