Learn More
Medchemexpress LLC Tamoxifen-d3 hydrochloride | 54965-24-1 | 99.0% | 410.99 | 1 MG
SDP
Catalog No. 5000352015
Encompass_Preferred
Tamoxifen-d3 hydrochloride is a deuterated labeled Tamoxifen that functions as an orally active, selective estrogen receptor modulator (SERM). It blocks estrogen action in breast cells and activates estrogen activity in other cells, such as bone, liver, and uterine cells. This compound is also a potent Hsp90 activator, enhancing its molecular chaperone ATPase activity. It potently inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 values of 0.1 μM and 1.8 μM, respectively. It activates autophagy and induces apoptosis, and can induce gene knockout of CreER(T2) transgenic mouse.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-52015 Supplier Medchemexpress LLC Supplier No. HY13757S1MG
Tamoxifen-d3 hydrochloride is a deuterated labeled Tamoxifen that functions as an orally active, selective estrogen receptor modulator (SERM). It blocks estrogen action in breast cells and activates estrogen activity in other cells, such as bone, liver, and uterine cells. This compound is also a potent Hsp90 activator, enhancing its molecular chaperone ATPase activity. It potently inhibits infectious EBOV Zaire and Marburg (MARV) with IC50 values of 0.1 μM and 1.8 μM, respectively. It activates autophagy and induces apoptosis, and can induce gene knockout of CreER(T2) transgenic mouse.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.