Learn More
Medchemexpress LLC Tamoxifen-d5 | 157698-32-3 | 99.99% | 376.55 | 1 MG
SDP
Catalog No. 5000350352
Encompass_Preferred
Tamoxifen-d5 is a deuterium labeled Tamoxifen. Tamoxifen (ICI 47699) is an orally active, selective estrogen receptor modulator (SERM). It is a potent Hsp90 activator and enhances Hsp90 molecular chaperone ATPase activity. Deuteration of drug molecules has gained attention for its potential to affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation
Catalog No. 50-003-50352 Supplier Medchemexpress LLC Supplier No. HY13757AS1MG
Tamoxifen-d5 is a deuterium labeled Tamoxifen. Tamoxifen (ICI 47699) is an orally active, selective estrogen receptor modulator (SERM). It is a potent Hsp90 activator and enhances Hsp90 molecular chaperone ATPase activity. Deuteration of drug molecules has gained attention for its potential to affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation