Learn More
Medi Dose Inc Eps Inc TAMPALERT 30ML VIALS 1000/PK
SDP
Catalog No. NC0931362
Encompass
TampAlerT 30 mL Vials (1,000 Vials). 30 mL Polyethylene Vial - Seals with a "Twist of the Wrist" No-Leak, Tamper-Evident Seal - Natural or White, Ultraviolet Inhibitant Polyethylene - USP Tight Child Resistant or Regular Screw Caps - In-House or Outpatient Dispensing Applications
Catalog No. NC0931362 Supplier Medi Dose Inc Eps Inc Supplier No. TA30
TampAlerT 30 mL Vials (1,000 Vials). 30 mL Polyethylene Vial - Seals with a "Twist of the Wrist" No-Leak, Tamper-Evident Seal - Natural or White, Ultraviolet Inhibitant Polyethylene - USP Tight Child Resistant or Regular Screw Caps - In-House or Outpatient Dispensing Applications