Learn More
GA International Tamper-Evident Cryogenic Labels 3.81" x 1.375" / 96.8mm x 34.9mm Rectangle White From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC1647362
Encompass_Preferred
Thermoplastic, waterproof cryogenic labels that will tear upon any attempt to remove them. Ideal for cryogenic vials, cryo boxes, and other containers that require tamper-evidence when stored in liquid nitrogen, dry ice, and freezers. Advance and practical design adds rigidity and strength to reduce accidental seal ruptures, yet destructs easily upon removal. Printable with thermal-transfer barcode printers.
Catalog No. NC1647362 Supplier GA International Supplier No. TELA5C11WH
Add to Cart
Add to Cart
Thermoplastic, waterproof cryogenic labels that will tear upon any attempt to remove them. Ideal for cryogenic vials, cryo boxes, and other containers that require tamper-evidence when stored in liquid nitrogen, dry ice, and freezers. Advance and practical design adds rigidity and strength to reduce accidental seal ruptures, yet destructs easily upon removal. Printable with thermal-transfer barcode printers.