Learn More
B BRAUN MED INC OEM TEC1000 Tamper Evident Syringe Cap - Red, 500 per case
Catalog No. NC2745461
Encompass_Preferred
-Used for capping Luer lock syringes. Tamper-evident provides improved quality control of prepared syringes.-Unique 10 per blister package protects sterility of unopened caps and aids in capping. 50 packs per case
This item is not returnable. View return policy
Catalog No. NC2745461 Supplier B BRAUN MED INC OEM Supplier No. 418004
-Used for capping Luer lock syringes. Tamper-evident provides improved quality control of prepared syringes.-Unique 10 per blister package protects sterility of unopened caps and aids in capping. 50 packs per case