Learn More
GA International Tamper-Evident Cryogenic Labels 0.75" x 1.8125" / 19.1mm x 46mm Rectangle White From -196°C to +110°C (-321°F to +230°F)
Catalog No. 5000571795
Encompass_Preferred
Thermoplastic, waterproof cryogenic labels that will tear upon any attempt to remove them. Ideal for cryogenic vials, cryo boxes, and other containers that require tamper-evidence when stored in liquid nitrogen, dry ice, and freezers. Advance and practical design adds rigidity and strength to reduce accidental seal ruptures, yet destructs easily upon removal. Printable with thermal-transfer barcode printers.
Catalog No. 50-005-71795 Supplier GA International Supplier No. TELA1NPC33WH
Thermoplastic, waterproof cryogenic labels that will tear upon any attempt to remove them. Ideal for cryogenic vials, cryo boxes, and other containers that require tamper-evidence when stored in liquid nitrogen, dry ice, and freezers. Advance and practical design adds rigidity and strength to reduce accidental seal ruptures, yet destructs easily upon removal. Printable with thermal-transfer barcode printers.