Learn More
Grainger NovaVision Inc. Tamper Evident Labels, Red, Polyester, 1-1/2 in. x 1 in., 250/Pk.
Catalog No. 502789057
Encompass_Preferred
These red tamper evident labels provide a secure, visible seal for files, envelopes, boxes, and other containers. Featuring a unique construction of flexible, spring-loaded strips connected between two security labels, they are highly sensitive to even the slightest tampering attempts, such as using a knife to slit a package. Once applied, the labels are virtually impossible to realign without leaving clear evidence of tampering. If either label is removed, it self-destructs to reveal a DOTS pattern.
  • Flexible design adapts to flat surfaces, corners, and unusual shapes
  • Aggressive adhesive is compatible with a wide range of surfaces
  • Sensitivity detects slitting and other minor tampering attempts
  • Strips spring out of alignment when tampered
  • Self-destructing feature leaves a visible DOTS pattern upon removal
  • Suitable for a wide range of container applications
Catalog No. 50-278-9057 Supplier Grainger Supplier No. 16G855
These red tamper evident labels provide a secure, visible seal for files, envelopes, boxes, and other containers. Featuring a unique construction of flexible, spring-loaded strips connected between two security labels, they are highly sensitive to even the slightest tampering attempts, such as using a knife to slit a package. Once applied, the labels are virtually impossible to realign without leaving clear evidence of tampering. If either label is removed, it self-destructs to reveal a DOTS pattern.
  • Flexible design adapts to flat surfaces, corners, and unusual shapes
  • Aggressive adhesive is compatible with a wide range of surfaces
  • Sensitivity detects slitting and other minor tampering attempts
  • Strips spring out of alignment when tampered
  • Self-destructing feature leaves a visible DOTS pattern upon removal
  • Suitable for a wide range of container applications