Learn More
Braintree Scientific Tamper Proof Dual Flow Regulator, (0.5-12 LPM)
SDP
Catalog No. 501954755
Encompass
Tamper-Proof Dual Flow RegulatorCompliant with AVMA Guidelines Factory preset to your specific cage size Easy operation with simple on/off valve 0.5 to 12 liter per minute (LPM) range Visual indicator ensures correct gas flow Tamper-Proof flow adjustment Connects directly to CO2 cylinder with CGA-320 fitting Includes 6 foot hose with quick disconnect fitting Simultaneously operate two different size cages **Sold for research purposes only and not for human use**
Catalog No. 50-195-4755 Supplier Braintree Scientific Supplier No. EP3300
Tamper-Proof Dual Flow RegulatorCompliant with AVMA Guidelines Factory preset to your specific cage size Easy operation with simple on/off valve 0.5 to 12 liter per minute (LPM) range Visual indicator ensures correct gas flow Tamper-Proof flow adjustment Connects directly to CO2 cylinder with CGA-320 fitting Includes 6 foot hose with quick disconnect fitting Simultaneously operate two different size cages **Sold for research purposes only and not for human use**