Learn More
MSC Osborn Counter Duster, Tampico Fill, 2 in Bristle Length, Wood Handle
Catalog No. 502836074
Encompass
This coarse counter duster is designed for heavier material removal and general factory or store dusting. It features a Tampico fill material firmly staple-set in premium sealed hardwood, with a handle that includes a hang-up hole for convenient storing.
  • Sturdy, light-weight brushing tool for removing dust accumulations from hard-to-reach places
  • Durable and long-lasting brush for dusting and chip removal
  • Can be used wet or dry and will not load
  • Tampico fill material
  • 2 inch bristle length
  • 8 inch brush area length
  • Straight wood handle
  • Gray bristle color
Catalog No. 50-283-6074 Supplier MSC Supplier No. 56008550
This coarse counter duster is designed for heavier material removal and general factory or store dusting. It features a Tampico fill material firmly staple-set in premium sealed hardwood, with a handle that includes a hang-up hole for convenient storing.
  • Sturdy, light-weight brushing tool for removing dust accumulations from hard-to-reach places
  • Durable and long-lasting brush for dusting and chip removal
  • Can be used wet or dry and will not load
  • Tampico fill material
  • 2 inch bristle length
  • 8 inch brush area length
  • Straight wood handle
  • Gray bristle color