Learn More
Electron Microscopy Sciences SecurTainer II™ Tamper Evident Specimen Containers, 20 ml
SDP
Catalog No. 501900256
Encompass_Preferred
SecurTainer II uses a different tamper evident concept compared to the SecurTainer I. No locking tab to push up is necessary when the tamper evident is used. Simply screw the cap on the container and the tamper evident sealing ring is automatically locking in place. When unscrewed, the ring is detached from the cap.
Container made of polypropylene is shatter resistant, especially designed for collection, transport and specimen storage.
Volume: 20 ml
Catalog No. 50-190-0256 Supplier Electron Microscopy Sciences Supplier No. 6423702
SecurTainer II uses a different tamper evident concept compared to the SecurTainer I. No locking tab to push up is necessary when the tamper evident is used. Simply screw the cap on the container and the tamper evident sealing ring is automatically locking in place. When unscrewed, the ring is detached from the cap.
Container made of polypropylene is shatter resistant, especially designed for collection, transport and specimen storage.
Volume: 20 ml