Learn More
Electron Microscopy Sciences SecurTainer I™ Tamper Evident Specimen Containers, 20 ml
SDP
Catalog No. 501900251
Encompass_Preferred
Tamper Evident Specimen polypropylene containers and closures are shatter resistant and feature a unique integrated leak resistant seal and ribbed lid for easy opening. SecurTrainer is a unique tamper evident to ensure no alter allow (SecurTainer™ can be used without using the tamper evident locking mechanism). High clarity to allow specimen to be viewed without opening. Both containers and caps are manufactured without the use of plasticizers or mold release agents. All material used are free from latex. All containers are 95kpa compliant. Non sterile, container and caps are packaged separately in bag of 100.
Volume: 20 ml
Catalog No. 50-190-0251 Supplier Electron Microscopy Sciences Supplier No. 6423602
Tamper Evident Specimen polypropylene containers and closures are shatter resistant and feature a unique integrated leak resistant seal and ribbed lid for easy opening. SecurTrainer is a unique tamper evident to ensure no alter allow (SecurTainer™ can be used without using the tamper evident locking mechanism). High clarity to allow specimen to be viewed without opening. Both containers and caps are manufactured without the use of plasticizers or mold release agents. All material used are free from latex. All containers are 95kpa compliant. Non sterile, container and caps are packaged separately in bag of 100.
Volume: 20 ml