Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Tamper-Proof Cryogenic Labels Rectangle, 2 x 1", White, 3" core
SDP

Catalog No. 501930434
Encompass_Preferred

These labels will withstand multiple freeze-thaw cycles, boiling water and autoclaving, dry ice, UV rays, Gamma irrradiation, most chemicals and solvents, and other harsh laboratory environments.

They are ideal for prolonged storage inside liquid nitrogen tanks, freezers and dry ice as well as for transportation of hazardous biological samples and other hazardous materials in dry ice containers, portable freezers and portable liquid nitrogen tanks.

Temperature range from -196 deg C to +110 deg C.
Size: 2 x 1"
Color: White
Core: 3"
Labels across: 1
Perforated: Yes

Catalog No. 50-193-0434 Supplier Electron Microscopy Sciences Supplier No. 7742806W
May include imposed supplier surcharges.
Add to Cart
Add to Cart

These labels will withstand multiple freeze-thaw cycles, boiling water and autoclaving, dry ice, UV rays, Gamma irrradiation, most chemicals and solvents, and other harsh laboratory environments.

They are ideal for prolonged storage inside liquid nitrogen tanks, freezers and dry ice as well as for transportation of hazardous biological samples and other hazardous materials in dry ice containers, portable freezers and portable liquid nitrogen tanks.

Temperature range from -196 deg C to +110 deg C.
Size: 2 x 1"
Color: White
Core: 3"
Labels across: 1
Perforated: Yes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.