Learn More
Abcam 5(6)-Carbo x ytetramethylrhodamine (TAMRA), 100MG
Supplier:  Abcam AB275361100MG
Encompass_Preferred
MW 860.9 Da, Purity >90%. Fluorescent dye that is used to label oligonucleotides at the 5'- or 3'-ends, or internally. It is also used for protein labeling where it can be added to the amino terminal or to lysine residues.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-512-94