Learn More
Medchemexpress LLC TAMRA alkyne, 5-isomer | 945928-17-6 | 96.6% | 467.52 g/mol | C28H25N3O4 | 10 MG
SDP
Catalog No. 5000281326
Encompass_Preferred
TAMRA alkyne, 5-isomer is a red-fluorescent tetramethylrhodamine derivative bearing a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC) labeling of biomolecules. Supplied as a solid small-molecule reagent, it is suitable for fluorescence labeling, enrichment, in-gel detection, and analytical identification.
Catalog No. 50-002-81326 Supplier Medchemexpress LLC Supplier No. HYW42308010MG
TAMRA alkyne, 5-isomer is a red-fluorescent tetramethylrhodamine derivative bearing a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC) labeling of biomolecules. Supplied as a solid small-molecule reagent, it is suitable for fluorescence labeling, enrichment, in-gel detection, and analytical identification.