Learn More
Medchemexpress LLC Tamra alkyne, 5-isomer | 945928-17-6 | 96.6% | 467.52 g/mol | C28H25N3O4 | 5 MG
SDP
Catalog No. 5000281269
Encompass_Preferred
TAMRA alkyne, 5-isomer is an alkyne-derivatized tetramethylrhodamine (TAMRA) fluorescent dye designed for click chemistry labeling. It permits efficient conjugation to azide- or DBCO/BCN-containing molecules and is commonly used for in-gel fluorescence detection, enrichment, and identification of labeled proteins.
Catalog No. 50-002-81269 Supplier Medchemexpress LLC Supplier No. HYW4230805MG
TAMRA alkyne, 5-isomer is an alkyne-derivatized tetramethylrhodamine (TAMRA) fluorescent dye designed for click chemistry labeling. It permits efficient conjugation to azide- or DBCO/BCN-containing molecules and is commonly used for in-gel fluorescence detection, enrichment, and identification of labeled proteins.