Learn More
Medchemexpress LLC TAMRA azide, 5-isomer | 825651-66-9 | C28H28N6O4 | 25 MG
SDP
Catalog No. 5000375993
Encompass_Preferred
TAMRA azide, 5-isomer is a click chemical containing azide groups that can be used as a linker for TAMRA. Its azide group enables copper-catalyzed click chemical reactions with alkynes, DBCO, and BCN. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups (Ex/Em = 555/580 nm).
  • Click chemistry reagent
  • Versatile linker
  • Reacts with alkynes, DBCO, and BCN
  • Copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Fluorescence properties: exhibits excitation/emission at 555/580 nm
Catalog No. 50-003-75993 Supplier Medchemexpress LLC Supplier No. 25MGHY151857
TAMRA azide, 5-isomer is a click chemical containing azide groups that can be used as a linker for TAMRA. Its azide group enables copper-catalyzed click chemical reactions with alkynes, DBCO, and BCN. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups (Ex/Em = 555/580 nm).
  • Click chemistry reagent
  • Versatile linker
  • Reacts with alkynes, DBCO, and BCN
  • Copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Fluorescence properties: exhibits excitation/emission at 555/580 nm