Learn More
Medchemexpress LLC Tamra azide, 5-isomer | 825651-66-9 | C28H28N6O4 | 50 MG
SDP
Catalog No. 5000372292
Encompass_Preferred
TAMRA azide, 5-isomer is a click chemical containing azide groups that can be used as a linker for TAMRA. The azide group enables copper-catalyzed click chemical reactions with alkynes, DBCO, and BCN. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups.
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups
  • Ex/Em = 555/580 nm
  • For research use only
Catalog No. 50-003-72292 Supplier Medchemexpress LLC Supplier No. HY15185750MG
TAMRA azide, 5-isomer is a click chemical containing azide groups that can be used as a linker for TAMRA. The azide group enables copper-catalyzed click chemical reactions with alkynes, DBCO, and BCN. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups.
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups
  • Ex/Em = 555/580 nm
  • For research use only