Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TAMRA azide, 6-isomer | 1192590-89-8 | 95.5% | 512.56 | 5 MG
SDP

Catalog No. 5000432011
Encompass_Preferred

TAMRA azide, 6-isomer is an isomer of TAMRA, a xanthene dye with orange emission. It is a solid, light brown to brown in appearance, primarily used as a fluorescent dye in various applications.

  • Molecular formula: C28H28N6O4
  • Molecular weight: 512.56
  • Store at 4°C, protect from light.
  • In solvent, store at -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility in DMSO: 50 mg/mL (97.55 mM)
  • Solubility in methanol: 5 mg/mL (9.75 mM)

Catalog No. 50-004-32011 Supplier Medchemexpress LLC Supplier No. HYD13065MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAMRA azide, 6-isomer is an isomer of TAMRA, a xanthene dye with orange emission. It is a solid, light brown to brown in appearance, primarily used as a fluorescent dye in various applications.

  • Molecular formula: C28H28N6O4
  • Molecular weight: 512.56
  • Store at 4°C, protect from light.
  • In solvent, store at -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility in DMSO: 50 mg/mL (97.55 mM)
  • Solubility in methanol: 5 mg/mL (9.75 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.