Learn More
Medchemexpress LLC Tamra azide, 6-isomer | 1192590-89-8 | MFCD29912896 | 95.5% | 512.56 | C28H28N6O4 | 10 MG
SDP
Catalog No. 5000297229
Encompass_Preferred
TAMRA azide, 6-isomer is an azide-functionalized 6-isomer of tetramethylrhodamine (TAMRA), a xanthene dye that emits in the orange region. It is supplied as a solid and is used for alkyne-azide click chemistry to label biomolecules for fluorescent detection and imaging.
  • Azide-functionalized 6-isomer for click chemistry.
  • Orange fluorescent xanthene dye for labeling and imaging.
  • High purity suitable for bioconjugation applications.
  • Available in multiple pack sizes for scale flexibility.
Catalog No. 50-002-97229 Supplier Medchemexpress LLC Supplier No. HYD130610MG
TAMRA azide, 6-isomer is an azide-functionalized 6-isomer of tetramethylrhodamine (TAMRA), a xanthene dye that emits in the orange region. It is supplied as a solid and is used for alkyne-azide click chemistry to label biomolecules for fluorescent detection and imaging.
  • Azide-functionalized 6-isomer for click chemistry.
  • Orange fluorescent xanthene dye for labeling and imaging.
  • High purity suitable for bioconjugation applications.
  • Available in multiple pack sizes for scale flexibility.