Learn More
Medchemexpress LLC Tamra azide, isomer 5 | 1006592-61-5 | 99.0% | 554.64 g/mol | C31H34N6O4 | 10mg
SDP
Catalog No. 5000268304
Encompass_Preferred
Tamra azide is a red fluorescent probe The azide group of Tamra azide can react with alkynes via copper-catalyzed click chemistry[1]
Catalog No. 50-002-68304 Supplier Medchemexpress LLC Supplier No. HYW80070510MG
Tamra azide is a red fluorescent probe The azide group of Tamra azide can react with alkynes via copper-catalyzed click chemistry[1]