Learn More
Medchemexpress LLC TAMRA-Azide-PEG-biotin | 1797415-74-7 | 99.0% | C57H79N11O14S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1409471MG
Encompass_Preferred
TAMRA-Azide-PEG-biotin is a dye derivative of TAMRA. It contains an azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Dye derivative of TAMRA.
  • Contains an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Solid, purple to purplish red appearance.
Catalog No. 50-003-55028