Learn More
CPC Scientific TAMRA-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH (trifluoroacetate salt) 0.1MG
SDP
Catalog No. 502108661
Encompass
beta-Amyloid (1-40) found to exhibit both neurotrophic and neurotoxic effects that depend on neuronal age and beta-protein concentration.ONE-LETTER SEQUENCE: TAMRA-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: C219H315N55O62S1MOLECULAR WEIGHT:4742.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802087-81-5], SYNONYMS: 5-TAMRA-Amyloid β-Protein (1-40)RESEARCH AREA: Alzheimer's DiseaseREFERENCES: B.A. Yankner et al., Science, 250, 279 (1990)
Catalog No. 50-210-8661 Supplier CPC Scientific Supplier No. AMYD012A
beta-Amyloid (1-40) found to exhibit both neurotrophic and neurotoxic effects that depend on neuronal age and beta-protein concentration.ONE-LETTER SEQUENCE: TAMRA-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVMOLECULAR FORMULA: C219H315N55O62S1MOLECULAR WEIGHT:4742.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1802087-81-5], SYNONYMS: 5-TAMRA-Amyloid β-Protein (1-40)RESEARCH AREA: Alzheimer's DiseaseREFERENCES: B.A. Yankner et al., Science, 250, 279 (1990)