Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tamra-PEG3-azide | 1228100-59-1 | 96.4% | 630.69 g/mol | C33H38N6O7 | 10MG
SDP

Catalog No. 5000237472
Encompass_Preferred

TAMRA-PEG3-Azide is a red-fluorescent TAMRA dye derivative bearing a three-unit polyethylene glycol (PEG3) spacer and a terminal azide functional group. It is intended for bioorthogonal conjugation to biomolecules via copper-catalyzed or strain-promoted azide-alkyne cycloaddition, offering a fluorescent label with improved solubility and spacer length.

  • Red-fluorescent dye with excitation/emission maxima 553/575 nm
  • Terminal azide enables CuAAC and SPAAC click chemistry
  • PEG3 spacer improves solubility and reduces steric interference
  • High purity (≈96.4%) in solid form
  • Soluble in methanol at about 12.5 mg/mL (19.82 mM); ultrasonic may be required
  • Storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month

Catalog No. 50-002-37472 Supplier Medchemexpress LLC Supplier No. HY12362910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAMRA-PEG3-Azide is a red-fluorescent TAMRA dye derivative bearing a three-unit polyethylene glycol (PEG3) spacer and a terminal azide functional group. It is intended for bioorthogonal conjugation to biomolecules via copper-catalyzed or strain-promoted azide-alkyne cycloaddition, offering a fluorescent label with improved solubility and spacer length.

  • Red-fluorescent dye with excitation/emission maxima 553/575 nm
  • Terminal azide enables CuAAC and SPAAC click chemistry
  • PEG3 spacer improves solubility and reduces steric interference
  • High purity (≈96.4%) in solid form
  • Soluble in methanol at about 12.5 mg/mL (19.82 mM); ultrasonic may be required
  • Storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.