Learn More
Medchemexpress LLC Tamra-peg3-azide | 1228100-59-1 | MFCD28334528 | 96.0% | 630.69 | C33H38N6O7 | 5MG
SDP
Catalog No. 5000236704
Encompass_Preferred
TAMRA-PEG3-Azide is a red-fluorescent dye derivative of 5-carboxytetramethylrhodamine bearing a three-unit polyethylene glycol (PEG3) linker and a terminal azide group. Supplied as a solid for research use, it enables bioconjugation through copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), allowing fluorescent labeling of alkyne-containing biomolecules.
Catalog No. 50-002-36704 Supplier Medchemexpress LLC Supplier No. HY1236295MG
TAMRA-PEG3-Azide is a red-fluorescent dye derivative of 5-carboxytetramethylrhodamine bearing a three-unit polyethylene glycol (PEG3) linker and a terminal azide group. Supplied as a solid for research use, it enables bioconjugation through copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC), allowing fluorescent labeling of alkyne-containing biomolecules.