Learn More
Medchemexpress LLC Tamra-peg4-acid | 1909223-02-4 | 95.3% | C36H43N3O10 | 25 MG
SDP
Catalog No. 5000352814
Encompass_Preferred
TAMRA-PEG4-acid is a dye derivative of TAMRA containing 3 PEG units. It is for research use only and not sold to patients.
  • Dye derivative of TAMRA
  • Contains 3 PEG units
  • Used in fluorescent dye applications
  • Related to labeling chemistry
  • A functional group type
  • Belongs to rhodamine dyes
Catalog No. 50-003-52814 Supplier Medchemexpress LLC Supplier No. 25MGHY140509
TAMRA-PEG4-acid is a dye derivative of TAMRA containing 3 PEG units. It is for research use only and not sold to patients.
  • Dye derivative of TAMRA
  • Contains 3 PEG units
  • Used in fluorescent dye applications
  • Related to labeling chemistry
  • A functional group type
  • Belongs to rhodamine dyes