Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TAMRA-PEG4-acid | 1909223-02-4 | 95.3% | C36H43N3O10 | 50 MG
SDP

Catalog No. 5000356663
Encompass_Preferred

TAMRA-PEG4-acid is a dye derivative of TAMRA containing 3 PEG units. This fluorescent dye is intended for research use only, suitable for laboratory chemicals and the manufacture of substances.

  • Target: Fluorescent dye
  • Molecular weight: 677.80
  • Solubility in DMSO: 100 mg/mL (147.54 mM)
  • Appearance: Liquid, brown to reddish brown

Catalog No. 50-003-56663 Supplier Medchemexpress LLC Supplier No. HY14050950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAMRA-PEG4-acid is a dye derivative of TAMRA containing 3 PEG units. This fluorescent dye is intended for research use only, suitable for laboratory chemicals and the manufacture of substances.

  • Target: Fluorescent dye
  • Molecular weight: 677.80
  • Solubility in DMSO: 100 mg/mL (147.54 mM)
  • Appearance: Liquid, brown to reddish brown

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.